missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 2.0mm (0.08 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 14761BQ

 View more versions of this product

Catalog No. AA14761BQ



Specifications

Specifications

Palladium wire
5cm
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
Odorless
palladium
23938
Wire
≈37.7g/m
2.0mm (0.08 in.) dia.
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
[Pd]
106.42
CHEBI:33363
99.95%

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents