missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Wire, 1mm (0.04 in.) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx.9.44g/m

Manufacturer:  Thermo Fisher Scientific 10280BW

 View more versions of this product

Catalog No. AA10280BW



Specifications

Specifications

Palladium
7440-05-3
12.02g/cm3 at 20°C
Pd
black
[Pd]
Odorless
23938
Wire, 1.0mm (0.04 in.) dia.
50cm
Silver gray wire
99.9% (Metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Silver to Gray

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black
CHEBI:33363
[Pd]
Documents