Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.5mm (0.02 in.) dia., Premion™, 99.99+% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium wire
CAS 7440-05-3
Name Note 0.5mm (0.02 in.) dia.
Purity Grade Premion™
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10961CB

  SDS
alfa aesar™
10961CB
250cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10961CD

  SDS
alfa aesar™
10961CD
1000cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium wire
TSCA TSCA
Name Note 0.5mm (0.02 in.) dia.
Purity Grade Premion™
Physical Form Wire
Assay ≥99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈2.36g/m
Quantity 250cm