Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.5mm (0.02 in.) dia., 99.9% (metals basis)

$160.08 - $160.08

Chemical Identifiers

Chemical Name or Material Palladium wire
CAS 7440-05-3
Name Note 0.5mm (0.02 in.) dia.
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10279BW

 
alfa aesar™
10279BW
50cm Each for $160.08

AA10279CB

 
alfa aesar™
10279CB
250cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10279CD

 
alfa aesar™
10279CD
1000cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium wire
TSCA TSCA
Name Note 0.5mm (0.02 in.) dia.
Physical Form Wire
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈2.36g/m
Quantity 50cm