missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.4mm (0.0159 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 46327G5

 View more versions of this product

Catalog No. AA46327G5



Specifications

Specifications

Palladium wire
5m
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
Odorless
palladium
23938
Wire
≈1.56g/m
0.4mm (0.0159 in.) dia., Annealed
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
[Pd]
106.42
CHEBI:33363
99.95%

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

RTECSNumber : RT3480500

TSCA : TSCA

SDS Documents