missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.25mm (0.01 in.) dia., Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 10962G1

 View more versions of this product

Catalog No. AA10962G1



Specifications

Specifications

Palladium wire
1m
7440-05-3
Pd
black
[Pd]
Odorless
23938
Wire
Premion™
≈0.6g/m
0.25mm (0.01 in.) dia.
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
99.99%

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents