missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.203mm (0.008 in.) dia., hard, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 45072G1

 View more versions of this product

Catalog No. AA45072G1



Specifications

Specifications

Palladium wire
0.203mm (0.008 in.) dia., Hard
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
[Pd]
106.42
CHEBI:33363
99.95%
1m
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
Odorless
palladium
23938
Wire

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents