missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Wire, 0.1mm (0.004 in.) dia., Premion™, 99.99% (Metals basis), Alfa Aesar™

Form: Approx.0.09g/m

Chemical Identifiers

CAS 7440-05-3
Physical Form Wire, 0.1mm (0.004 in.) dia.
Assay Percent Range 99.99% (Metals basis)
MDL Number MFCD00011167
Chemical Name or Material Palladium
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10963G5 SDS Alfa Aesar
10963G5
5m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10963G9 SDS Alfa Aesar
10963G9
25m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7440-05-3
99.99% (Metals basis)
Palladium
Wire, 0.1mm (0.004 in.) dia.
MFCD00011167

Specifications

5m
Silver gray wire
99.99% (Metals basis)
Odorless
Palladium
7440-05-3
Wire, 0.1mm (0.004 in.) dia.
MFCD00011167
Silver-gray