Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 0.02mm (0.0008 in.) dia., 99.9% (metals basis) 1m

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 0.02mm (0.0008 in.) dia., Temper: as drawn
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47134G1

 
alfa aesar™
47134G1
1m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 0.02mm (0.0008 in.) dia., Temper: as drawn
Physical Form Wire
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Color Gold
RTECS Number RT3480500
Weight ≈0.004g/m
Quantity 1m