Please login to your online account to display your discounted pricing

Palladium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLPD32Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLPD3-2Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Health Hazard 1 Causes skin irritation, serious eye irritation. May cause respiratory irritation
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Solvent or Matrix 10% HCl
DOT Information Hazardous
Recommended Storage Ambient
Quantity 125mL
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
10% HCl    1000μg/mL    PLPD3-2Y, -2X    10,000    PLPD3-3Y, -3X
Chemical Name or Material Palladium Standard
CAS 7732-18-5
CAS Max % 89.9
CAS Min % 10.0
CAS Max % 0.1
CAS 7440-05-3
CAS Min % 0.1
CAS Max % 10.0
CAS Min % 89.9
CAS 7647-01-0

Specifications

Chemical Name or Material Palladium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pd1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation. May cause respiratory irritation

Ambient