Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40418KG

 
alfa aesar™
40418KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40418KJ

 
alfa aesar
40418KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Palladium slug
TSCA TSCA
Name Note 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Physical Form Slug
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈2.41g/slug
Quantity 1 piece

  • Evaporation source, hydrogen getter, jewelry production, electrical contacts for semiconductors