Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium Silver foil, 0.127mm (0.005 in.) thick, 99.9% (metals basis excluding Pt)

Chemical Identifiers

Chemical Name or Material Palladium Silver foil
Name Note 0.127mm (0.005 in.) thick
MDL Number MFCD02091738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12575FF

  SDS
alfa aesar™
12575FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12575FI

  SDS
alfa aesar™
12575FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12575GH

  SDS
alfa aesar™
12575GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium Silver foil
TSCA TSCA
Name Note 0.127mm (0.005 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (metals basis excluding Pt)
MDL Number MFCD02091738
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Pd:Ag; 75:25 wt%
Weight ≈0.92g/25x25mm
Quantity 25x25mm