missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium Silver foil, 0.127mm (0.005 in.) thick, 99.9% (metals basis excluding Pt)

Manufacturer:  Alfa Aesar™ 12575FI

 View more versions of this product

Catalog No. AA12575FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pd:Ag; 75:25 wt%
≈0.92g/25x25mm
0.127mm (0.005 in.) thick
MFCD02091738
Foil
Palladium Silver foil
50 x 50mm
(metals basis excluding Pt)
Odorless
99.9%
Documents