Alfa Aesar™ Palladium Silver foil, 0.10mm (0.004 in.) thick, 99.9% (metals basis excluding Pt)
Manufacturer:  Alfa Aesar™ 44731FF
 View more versions of this product
Catalog No. AA44731FF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Pd:Ag; 75:25 wt%
≈0.72g/25x25mm
0.10mm (0.004 in.) thick
MFCD02091738
99.9%
Palladium Silver foil
25 x 25mm
(metals basis excluding Pt)
Foil
Documents