missing translation for 'onlineSavingsMsg'
Learn More

Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)

Pd:Ag; 75:25 wt%

$263.03 - $263.03

Chemical Identifiers

Molecular Formula Pd:Ag; 75:25 wt%
MDL Number MFCD02091738
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42187FF
View Documents
Thermo Scientific Chemicals
42187FF
25 x 25 mm
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

Pd:Ag; 75:25 wt%
MFCD02091738

Specifications

Tube
MFCD02091738
99.9%
Pd:Ag; 75:25 wt%
25 x 25 mm
Palladium Silver foil
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only