Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium rod, 6.35mm (0.25 in.) dia., 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium rod
CAS 7440-05-3
Name Note 6.35mm (0.25 in.) dia.
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12557DH

 
alfa aesar™
12557DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12557DM

 
alfa aesar™
12557DM
100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium rod
TSCA TSCA
Name Note 6.35mm (0.25 in.) dia.
Physical Form Rod
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈3.80g/cm
Quantity 25mm