missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium rod, 6.35mm (0.25 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 12557DM

 View more versions of this product

Catalog No. AA12557DM



Specifications

Specifications

Palladium rod
100mm
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
Odorless
23938
Rod
≈3.80g/cm
6.35mm (0.25 in.) dia.
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
99.95%

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents