missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium powder, spherical, APS 0.5-1.7 micron, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4371506

 View more versions of this product

Catalog No. AA4371506



Specifications

Specifications

Palladium powder
(metals basis)
MFCD00011167
black
[Pd]
106.42
CHEBI:33363
99.95%
Odorless
0.7 to 2.0m2/g
7440-05-3
Pd
UN3089
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Sperical Powder
Tap density: 2.4 to 5.0g/cm3
APS 0.5 to 1.7 Micron
5g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : TSCA

Documents