Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon paste, A102023-5, Alfa Aesar™

Form: 300m2/g

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 1% on alumina powder
Physical Form Powder
MDL Number MFCD03613602
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830006

 
alfa aesar
3830006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830014

 
alfa aesar
3830014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830022

 
alfa aesar
3830022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Color Light gray
Chemical Name or Material Palladium
Name Note 1% on alumina powder
Physical Form Powder
MDL Number MFCD03613602
Odor Odorless
Appearance Light gray powder