Please login to your online account to display your discounted pricing

Palladium Powder, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 5% on barium sulfate powder
Physical Form Powder
MDL Number MFCD03613605
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1172214

  SDS
alfa aesar
1172214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1172222

  SDS
alfa aesar
1172222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1172288

  SDS
alfa aesar
1172288
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Color Gray-black
Physical Form Powder
Appearance grayish black powder
Odor Odorless
MDL Number MFCD03613605
Chemical Name or Material Palladium
Name Note 5% on barium sulfate powder