missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Powder, Alfa Aesar™

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD03427451
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1172106
View Documents
Thermo Fisher Scientific
1172106
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1172114
View Documents
Thermo Fisher Scientific
1172114
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1172122
View Documents
Thermo Fisher Scientific
1172122
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03427451
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
CHEBI:33363
[Pd]

Specifications

5g
gray powder
MFCD03427451
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Gray
7440-05-3
Pd
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
[Pd]
Odorless
23938
Powder
Palladium, 5% on barium carbonate powder
Safety and Handling

Safety and Handling

Avoid contact with skin and eyes.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : DOT Class: 6.1, Packaging Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures