missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Powder, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1172114

 View more versions of this product

Catalog No. AA1172114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Palladium
5% on barium carbonate powder
gray powder
MFCD03427451
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Gray
25g
7440-05-3
Pd
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
[Pd]
Odorless
23938
Powder

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03427451
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

Avoid contact with skin and eyes.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : DOT Class: 6.1, Packaging Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents