missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Powder, -200 mesh, 99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1206806

 View more versions of this product

Catalog No. AA1206806



Specifications

Specifications

Palladium
7440-05-3
99.95% (Metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Gray to Black
5g
Dark gray to black powder
Pd
black
[Pd]
Odorless
23938
Powder

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

H228 : Flammable solid

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-115-6

Documents