missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14432AE

 View more versions of this product

Catalog No. AA14432AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Palladium
Liquid
MFCD03613603
[Pd]
106.42
CHEBI:33363
Characteristic
7440-05-3
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Liquid
100mL

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
CHEBI:33363
[Pd]
Pd
MFCD03613603
23938
palladium
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

Documents