Learn More
Palladium, plasma standard solution, Specpure™, Pd 1000μg/ml, Alfa Aesar™
Manufacturer: Alfa Aesar 13833AE
Specifications
Palladium, plasma standard solution, Specpure™ | |
MFCD03613603 | |
Pd 1000μg/ml | |
100mL |
Pd in 20% HCl | |
UN1789 | |
Liquid |
Chemical Identifiers
Pd in 20% HCl | |
MFCD03613603 |
Safety and Handling
GHS H Statement
H314-H335;
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
TSCA : TSCA