Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) trimethylacetate

Chemical Identifiers

Chemical Name or Material Palladium(II) trimethylacetate
CAS 106224-36-6
Linear Formula [(CH3)3CCO2]2Pd
Formula Weight 308.67
MDL Number MFCD16038138
Synonym Palladium(II) pivalate; 2,2-Dimethylpropionic acid palladium(II) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4668503

 
alfa aesar™
4668503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 308.67
Melting Point 230° to 232°C
Chemical Name or Material Palladium(II) trimethylacetate
Physical Form Solid
Linear Formula [(CH3)3CCO2]2Pd
MDL Number MFCD16038138
CAS 106224-36-6
Synonym Palladium(II) pivalate; 2,2-Dimethylpropionic acid palladium(II) salt
Color Orange
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.