Learn More
Palladium(II) trimethylacetate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04668503
Description
Palladium(II) trimethylacetate is used as a catalyst for aerobic cyclization of acrylic acid with alkenes. It is utilized in the benzylation of heterocycles with benzyl chloride and direct arylation reactions. Furthermore, it is used in regioselective oxidative arene cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium(II) trimethylacetate | |
Orange | |
Solid | |
C10H18O4Pd | |
MFCD16038138 | |
Soluble in ethyl acetate,methanol,chloroform,tetrahydrofuran and 1,4-dioxane. Insoluble in water. | |
CC(C)(C)C(=O)[O-].CC(C)(C)C(=O)[O-].[Pd+2] | |
308.67 | |
308.67 |
230°C to 232°C | |
106224-36-6 | |
1 g | |
[(CH3)3CCO2]2Pd | |
palladium pivalate, palladium ii pivalate, palladium ii trimethylacetate, acmc-20ajzo, palladium 11 pivalate, palladium 2+ dipivalate, dipivalic acid palladium ii salt, palladium 2+ ion dipivalate, palladium 2+ bis 2,2-dimethylpropanoate | |
PAGZTSLSNQZYEV-UHFFFAOYSA-L | |
2,2-dimethylpropanoate;palladium(2+) | |
24905427 |
Chemical Identifiers
106224-36-6 | |
308.67 | |
PAGZTSLSNQZYEV-UHFFFAOYSA-L | |
24905427 | |
CC(C)(C)C(=O)[O-].CC(C)(C)C(=O)[O-].[Pd+2] |
C10H18O4Pd | |
MFCD16038138 | |
palladium pivalate, palladium ii pivalate, palladium ii trimethylacetate, acmc-20ajzo, palladium 11 pivalate, palladium 2+ dipivalate, dipivalic acid palladium ii salt, palladium 2+ ion dipivalate, palladium 2+ bis 2,2-dimethylpropanoate | |
2,2-dimethylpropanoate;palladium(2+) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only