Learn More
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03944803
Description
Palladium(II) trifluoroacetate is used in the preparation of allyl-palladium complexes. It acts as a catalyst used in the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes. Further, it is involved in the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates in order to prepare keto alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium(II) trifluoroacetate | |
Brown | |
Powder | |
97% | |
MFCD00013204 | |
Soluble in diethyl ether and acetone. Insoluble in benzene,chloroform and trifluoroacetic acid. | |
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F | |
332.45 | |
332.44 | |
Brown powder |
210°C (decomposition) | |
42196-31-6 | |
1 g | |
C4F6O4Pd | |
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d | |
PBDBXAQKXCXZCJ-UHFFFAOYSA-L | |
palladium(2+) ditrifluoroacetate | |
2733347 | |
Hygroscopic |
Chemical Identifiers
42196-31-6 | |
332.45 | |
PBDBXAQKXCXZCJ-UHFFFAOYSA-L | |
2733347 |
C4F6O4Pd | |
MFCD00013204 | |
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d | |
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only