missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) sulfide, 99% (metals basis), Pd 72% min

Manufacturer:  Alfa Aesar™ 1267403

 View more versions of this product

Catalog No. AA1267403



Specifications

Specifications

Palladium(II) sulfide
(metals basis)
MFCD00016285
NRUVOKMCGYWODZ-UHFFFAOYSA-N
sulfanylidenepalladium
82926
Crystalline
Pd 72% min.
Sulphurous
1g
12125-22-3
PdS
palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
S=[Pd]
138.48
138.46
99%
6.60g/mL
950°C (decomposition)

Chemical Identifiers

12125-22-3
138.48
NRUVOKMCGYWODZ-UHFFFAOYSA-N
82926
S=[Pd]
PdS
MFCD00016285
palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
sulfanylidenepalladium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents