Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) sulfide, 99% (metals basis), Pd 72% min

Chemical Identifiers

Chemical Name or Material Palladium(II) sulfide
CAS 12125-22-3
Formula Weight 138.46
MDL Number MFCD00016285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1267403

 
alfa aesar™
1267403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1267406

 
alfa aesar™
1267406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.46
Physical Form Crystalline
Melting Point 950°C (dec.)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Pd 72% min.
Chemical Name or Material Palladium(II) sulfide
TSCA TSCA
Odor Sulphurous
Assay 99%
Assay Percent Notes (metals basis)
MDL Number MFCD00016285
CAS 12125-22-3
Density 6.60g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.