missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) sulfate, solution, Pd 6% w/w (cont. Pd)

Manufacturer:  Alfa Aesar™ 12622NB

 View more versions of this product

Catalog No. AA12622NB



Specifications

Specifications

Palladium(II) sulfate
13566-03-5
PdSO4
UN3264
palladium ii sulfate, palladium sulfate, palladium 2+ sulfate, palladium sulphate, palladous sulfate, palladium monosulfate, palladium sulfate pdso4, sulfuric acid, palladium salt, acmc-20ajzn, pdso4
[O-]S(=O)(=O)[O-].[Pd+2]
202.476
202.48 (Anhydrous)
Red to Brown
Solution
O4PdS
MFCD00011173
Pd 6% w/w (cont. Pd)
RFLFDJSIZCCYIP-UHFFFAOYSA-L
palladium(2+);sulfate
166846
Liquid
5g

Chemical Identifiers

13566-03-5
202.476
RFLFDJSIZCCYIP-UHFFFAOYSA-L
166846
[O-]S(=O)(=O)[O-].[Pd+2]
O4PdS
MFCD00011173
palladium ii sulfate, palladium sulfate, palladium 2+ sulfate, palladium sulphate, palladous sulfate, palladium monosulfate, palladium sulfate pdso4, sulfuric acid, palladium salt, acmc-20ajzn, pdso4
palladium(2+);sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H317
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents