missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 4369806

 View more versions of this product

Catalog No. AA4369806



Specifications

Specifications

Palladium (II) sulfate dihydrate
(metals basis)
PdSO4·2H2O
UN3260
TWIRRPLUAGEFNJ-UHFFFAOYSA-L
palladium(2+);sulfate;dihydrate
22042048
Powder
Premion™
Brown to Red
13444-98-9
H4O6PdS
MFCD00011173
palladium 2+ dihydrate sulfate
O.O.[O-]S(=O)(=O)[O-].[Pd+2]
238.506
238.51 (202.48 Anhydrous)
99.995%
Hygroscopic
5g

Chemical Identifiers

13444-98-9
238.506
TWIRRPLUAGEFNJ-UHFFFAOYSA-L
22042048
O.O.[O-]S(=O)(=O)[O-].[Pd+2]
H4O6PdS
MFCD00011173
palladium 2+ dihydrate sulfate
palladium(2+);sulfate;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents