Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) sulfate dihydrate, Premion™, 99.95% (metals basis), Pd 44.1% min

Chemical Identifiers

Chemical Name or Material Palladium(II) sulfate dihydrate
CAS 13444-98-9
Purity Grade Premion™
Linear Formula PdSO4·2H2O
Formula Weight 238.51 (202.48 Anhydrous)
MDL Number MFCD00011173
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1216501

 
alfa aesar™
1216501
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1216503

 
alfa aesar™
1216503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1216506

 
alfa aesar™
1216506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.51 (202.48 Anhydrous)
Chemical Name or Material Palladium(II) sulfate dihydrate
TSCA TSCA
Purity Grade Premion™
Physical Form Crystalline
Assay 99.95%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula PdSO4·2H2O
MDL Number MFCD00011173
CAS 13444-98-9
UN Number UN3260
Color Red to brown
Quantity 0.25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Pd 44.1% min.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.