Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) oxide monohydrate, 99.9% (metals basis), Pd 73% min

Chemical Identifiers

Chemical Name or Material Palladium(II) oxide monohydrate
CAS 64109-12-2
Formula Weight 140.41 (122.40 Anhydrous)
MDL Number MFCD00011172
Merck Index 14,6993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1104104

  SDS
alfa aesar™
1104104
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1104109

  SDS
alfa aesar™
1104109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.41 (122.40 Anhydrous)
CAS 64109-12-2
Merck Index 14,6993
Chemical Name or Material Palladium(II) oxide monohydrate
MDL Number MFCD00011172
Assay 99.9%
Concentration Pd 73% min.
Quantity 2g
TSCA TSCA
Physical Form Powder
Assay Percent Notes (metals basis)