missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min

Catalytic reduction, in organic synthesis

Manufacturer:  Alfa Aesar™ 1104003

 View more versions of this product

Catalog No. AA1104003



Specifications

Specifications

Palladium(II) oxide
1314-08-5
MFCD00011172
palladium ii oxide, palladium oxide pdo, palladium ii oxide, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
oxopalladium
5083724
Powder
Pd 85% min.
−22 Mesh
Brown
1g
Anhydrous
OPd
14,6993
Insoluble in water, acids; Slightly soluble in aqua regia
O=[Pd]
122.419
122.4
99.9%
8.70g/mL
Odorless
870°C

Chemical Identifiers

1314-08-5
122.419
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
5083724
O=[Pd]
OPd
MFCD00011172
palladium ii oxide, palladium oxide pdo, palladium ii oxide, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
oxopalladium
Safety and Handling

Safety and Handling

RTECSNumber : RT3592900

TSCA : TSCA

Documents