Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min

Catalytic reduction, in organic synthesis

$198.89 - $198.89

Chemical Identifiers

Chemical Name or Material Palladium(II) oxide
CAS 1314-08-5
Name Note Anhydrous
Formula Weight 122.4
MDL Number MFCD00011172
Merck Index 14,6993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1104003

  SDS
alfa aesar™
1104003
1g Each for $198.89

AA1104006

  SDS
alfa aesar™
1104006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium(II) oxide
Name Note Anhydrous
CAS 1314-08-5
RTECS Number RT3592900
TSCA TSCA
Physical Form Powder
Assay 99.9%
Formula Weight 122.4
MDL Number MFCD00011172
Merck Index 14,6993
Solubility Information Insoluble in water, acids; Slightly soluble in aqua regia
Concentration Pd 85% min.
Density 8.70g/mL
Mesh Size −22 Mesh
Odor Odorless
Color Brown
Melting Point 870°C
Quantity 1g