Please login to your online account to display your discounted pricing

Palladium(II) oxide, 86.8-87.1% Pd, ACROS Organics™

$165.03 - $165.03

Chemical Identifiers

Chemical Name or Material Palladium(II) oxide
Name Note 86.8-87.1% Pd
CAS 1314-08-5
Assay 86.8 to 87.1% (Pd)
Molecular Formula OPd
Linear Formula PdO
Formula Weight 122.4
MDL Number MFCD00011172
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195160010

  SDS
thermo fisher scientific
195160010
Glass bottle 1g Each for $165.03

AC195160050

  SDS
thermo fisher scientific
195160050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium(II) oxide
Name Note 86.8-87.1% Pd
CAS 1314-08-5
EINECS Number 215-218-3
TSCA TSCA
Physical Form Powder
Assay 86.8 to 87.1% (Pd)
Molecular Formula OPd
Linear Formula PdO
Formula Weight 122.4
MDL Number MFCD00011172
Merck Index 15, 7094
Density 8.7
Packaging Glass bottle
Color Gray to black
Quantity 1g