Learn More
Alfa Aesar™ Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd)
Manufacturer: Alfa Aesar™ 12621NH
Specifications
Palladium(II) nitrate | |
10102-05-3 | |
MFCD00011169 | |
Pd 10% w/w (cont. Pd) | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
palladium(2+);dinitrate | |
24932 | |
Liquid | |
1g |
Solution | |
N2O6Pd | |
UN3264 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium ii nitrate, palladium 2+ dinitrate, palladiumdinitrate | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pd+2] | |
230.428 | |
230.43 | |
Brown to Orange |
Chemical Identifiers
10102-05-3 | |
230.428 | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
24932 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pd+2] |
N2O6Pd | |
MFCD00011169 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium ii nitrate, palladium 2+ dinitrate, palladiumdinitrate | |
palladium(2+);dinitrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
RTECSNumber : QV0560000
TSCA : TSCA