Please login to your online account to display your discounted pricing

Palladium(II) nitrate hydrate, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC389910050

 View more versions of this product

Catalog No. AC389910050

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Palladium(II) nitrate hydrate
CAS 207596-32-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May intensify fire; oxidizer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
CAS 10102-05-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Physical Form Crystals
Molecular Formula N2O6Pd·xH2O
Linear Formula Pd(NO3)2·xH2O
Formula Weight 230.43
MDL Number MFCD00149818
Packaging Glass bottle
Palladium (Pd) 39 to 46%
Color Dark brown-black
Quantity 5g

Specifications

Chemical Name or Material Palladium(II) nitrate hydrate
CAS 207596-32-5
CAS 10102-05-3
Molecular Formula N2O6Pd·xH2O
Linear Formula Pd(NO3)2·xH2O
Formula Weight 230.43
MDL Number MFCD00149818
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May intensify fire; oxidizer.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents