Please login to your online account to display your discounted pricing

Palladium(II) nitrate hydrate, 99.9% (metals basis), Pd 39% min, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Palladium(II) Nitrate Hydrate
CAS 10102-05-3
Physical Form Crystals
Assay Percent Range 99.8% (Metals Basis)
Molecular Formula N2O6Pd·xH2O
Formula Weight 230.43
MDL Number MFCD00011169
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1103518

  SDS
alfa aesar
1103518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103509

  SDS
alfa aesar
1103509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium(II) Nitrate Hydrate
CAS 10102-05-3
Health Hazard 1 H272-H400-H410-H315-H319-H317: May intensify fire; oxidizer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
DOT Information DOT Class: 5.1, Packaging Group: III
EINECS Number 233-265-8
Physical Form Crystals
Assay Percent Range 99.8% (Metals Basis)
Molecular Formula N2O6Pd·xH2O
Appearance Red-Brown Crystals
Formula Weight 230.43
MDL Number MFCD00011169
Color Red-brown
Quantity 50g
Safety and Handling

H272-H400-H410-H315-H319-H317: May intensify fire; oxidizer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.