missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium(II) nitrate hydrate, 99.9% (metals basis), Pd 39% min, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1103504

 View more versions of this product

Catalog No. AA1103504



Specifications

Specifications

Palladium(II) nitrate hydrate
(metals basis), Pd 39% min
Pd(NO3)2·xH2O
UN1477
hydrogen tetranitropalladate ii
GPNDARIEYHPYAY-UHFFFAOYSA-N
palladium(2+);dinitrate
24932
Crystalline
Hygroscopic
2g
10102-05-3
N2O6Pd
MFCD00011169
14,6992
Soluble in water with turbidity, precipitating a brown basic salt. Completely soluble in dilute HNO3
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pd+2]
230.428
230.43 (Anhydrous)
99.8%
Brown to Red

Chemical Identifiers

10102-05-3
230.428
GPNDARIEYHPYAY-UHFFFAOYSA-N
24932
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pd+2]
N2O6Pd
MFCD00011169
hydrogen tetranitropalladate ii
palladium(2+);dinitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H317;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P220-P261-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : QV0560000

TSCA : TSCA

Documents