Please login to your online account to display your discounted pricing

Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min, Alfa Aesar™

$151.47 - $151.47

Chemical Identifiers

Chemical Name or Material Palladium(II) nitrate hydrate
CAS 10102-05-3
Molecular Formula N2O6Pd·xH2O
Linear Formula Pd(NO3)2·xH2O
Formula Weight 230.43 (Anhydrous)
MDL Number MFCD00011169
UN Number UN1477
Merck Index 14,6992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1103504

 
alfa aesar
1103504
Each for $151.47
Description & Specifications

Specifications

Formula Weight 230.43 (Anhydrous)
Quantity 2g
Chemical Name or Material Palladium(II) nitrate hydrate
TSCA TSCA
Physical Form Crystalline
Assay 99.8%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis), Pd 39% min
Molecular Formula N2O6Pd·xH2O
Linear Formula Pd(NO3)2·xH2O
MDL Number MFCD00011169
CAS 10102-05-3
UN Number UN1477
Merck Index 14,6992
Color Red to Brown
Solubility Information Soluble in water with turbidity, precipitating a brown basic salt. Completely soluble in dilute HNO3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319-H317;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P220-P261-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
RTECS Number QV0560000
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P261-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H317;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.