missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) chloride, solution, Pd 9.0-11.0% w/w (cont. Pd)

Manufacturer:  Alfa Aesar™ 12623NB

 View more versions of this product

Catalog No. AA12623NB



Specifications

Specifications

Palladium(II) chloride
7647-10-1
MFCD00003558
Pd 9.0 to 11.0% w/w (cont. Pd)
PIBWKRNGBLPSSY-UHFFFAOYSA-L
dichloropalladium
24290
177.31
Acidic
5g
Solution
Cl2Pd
UN3264
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium ii chloride, palladium chloride pdcl2, palladium ii chloride, enplate activator 440
Cl[Pd]Cl
177.32
CHEBI:53434
Liquid
Brown

Chemical Identifiers

7647-10-1
177.32
PIBWKRNGBLPSSY-UHFFFAOYSA-L
24290
dichloropalladium
Cl2Pd
MFCD00003558
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium ii chloride, palladium chloride pdcl2, palladium ii chloride, enplate activator 440
CHEBI:53434
Cl[Pd]Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents