missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min

Manufacturer:  Alfa Aesar™ 4369706

 View more versions of this product

Catalog No. AA4369706



Specifications

Specifications

Palladium (II) bromide
(metals basis)
MFCD00011170
INIOZDBICVTGEO-UHFFFAOYSA-L
dibromopalladium
83469
Powder
Premion™
Black
13444-94-5
Br2Pd
palladium 2+ dibromide
Br[Pd]Br
266.228
266.22
99.998%
Pd 39.5% min
5g

Chemical Identifiers

13444-94-5
266.228
INIOZDBICVTGEO-UHFFFAOYSA-L
83469
Br[Pd]Br
Br2Pd
MFCD00011170
palladium 2+ dibromide
dibromopalladium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents