Please login to your online account to display your discounted pricing

Palladium(II) bromide, 99.99% (metals basis), Pd 39.5% min., Alfa Aesar™ Premion™

Chemical Identifiers

Chemical Name or Material Palladium hydroxide
CAS 12135-22-7
Physical Form Moist Powder
Assay Percent Range 99.99% (metal basis)
Molecular Formula PdBr2
Formula Weight 266.22
MDL Number MFCD00064599
Synonym Pearlman's Catalyst
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1187804

 
alfa aesar
1187804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1187809

 
alfa aesar
1187809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Moist Powder
Formula Weight 266.22
Quantity 2g
Color Black
Chemical Name or Material Palladium hydroxide
EINECS Number 235-219-2
Odor Odorless
Assay Percent Range 99.99% (metal basis)
Molecular Formula PdBr2
Appearance Black moist powder
MDL Number MFCD00064599
CAS 12135-22-7
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Synonym Pearlman's Catalyst