missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium(II) Acetate Trimer 98.0+%, TCI America™

Supplier:  TCI America P21061G

 View more versions of this product

Catalog No. P21061G



Description

Description

Specifications

Specifications

Palladium(II) Acetate Trimer
C12H24O12Pd3
Hexakis(acetato)tripalladium(II), Tripalladium(II) Hexaacetate
CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pd].[Pd].[Pd]
679.572
673.52
Crystalline Powder
53189-26-7
1 g
FUKTVTHLBVPABW-UHFFFAOYSA-N
acetic acid;palladium
85470766
≥98.0% (T)

Chemical Identifiers

53189-26-7
679.572
Hexakis(acetato)tripalladium(II), Tripalladium(II) Hexaacetate
acetic acid;palladium
C12H24O12Pd3
FUKTVTHLBVPABW-UHFFFAOYSA-N
85470766
CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pd].[Pd].[Pd]
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0693

TSCA : No

SDS
Documents

Documents

Promotions

Promotions