Please login to your online account to display your discounted pricing

Palladium(II) acetate, trimer, Pd 45.9-48.4%, Alfa Aesar™

$110.74 - $1,298.82

Chemical Identifiers

Chemical Name or Material Palladium(II) acetate
CAS 3375-31-3
Physical Form Various Forms In Red-(Powder/Flake/Crystalline/Beads)
Assay Percent Range 45.9 to 48.4%
Molecular Formula C12H18O12Pd3
Formula Weight 673.46
MDL Number MFCD00012453
Synonym Acetic acid palladium(II) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1051614

  SDS
alfa aesar
1051614
25g Each for $1,298.82

AA1051603

  SDS
alfa aesar
1051603
1g Each for $110.74

AA1051606

  SDS
alfa aesar
1051606
5g Each for $405.32
Description & Specifications

Specifications

Chemical Name or Material Palladium(II) acetate
CAS 3375-31-3
Health Hazard 1 H318-H317-H413: Causes serious eye damage. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life.
Health Hazard 2 Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
EINECS Number 222-164-4
Physical Form Various Forms In Red-(Powder/Flake/Crystalline/Beads)
Assay Percent Range 45.9 to 48.4%
Molecular Formula C12H18O12Pd3
Appearance Various forms in red-brown color (powder/flake/crystalline/beads)
Formula Weight 673.46
MDL Number MFCD00012453
Synonym Acetic acid palladium(II) salt
Color Red-brown
Quantity 25g
Safety and Handling

H318-H317-H413: Causes serious eye damage. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life.

Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.