missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium(II) acetate, 99.9%, Acros Organics

Manufacturer:  Acros Organics 441390050

 View more versions of this product

Catalog No. AC441390050



Description

Description

For ultra trace analysis
Specifications

Specifications

Palladium(II) Acetate
99.9%
(CH3CO2)2Pd
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium(2+);diacetate
167845
99.9%
Glass bottle
205.0°C
3375-31-3
C4H6O4Pd
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, palladium ii acetate, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
CC(=O)[O-].CC(=O)[O-].[Pd+2]
224.508
224.51
Trace Metal Basis
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols
5g

Chemical Identifiers

3375-31-3
224.508
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, palladium ii acetate, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
palladium(2+);diacetate
C4H6O4Pd
YJVFFLUZDVXJQI-UHFFFAOYSA-L
167845
CC(=O)[O-].CC(=O)[O-].[Pd+2]
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents