missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium(II) acetate, 47.5% Pd, ACROS Organics™

Manufacturer:  Acros Organics AC195180010

 View more versions of this product

Catalog No. AC195180010



Specifications

Specifications

Palladium(II) acetate
3375-31-3
(CH3CO2)2Pd
bis acetato palladium
CC(=O)[O-].CC(=O)[O-].[Pd+2]
224.508
224.51
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols
1g
47.5% Pd
C4H6O4Pd
MFCD00012453
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium(2+);diacetate
167845
Glass bottle
205.0°C

Chemical Identifiers

3375-31-3
224.508
YJVFFLUZDVXJQI-UHFFFAOYSA-L
167845
CC(=O)[O-].CC(=O)[O-].[Pd+2]
C4H6O4Pd
MFCD00012453
bis acetato palladium
palladium(2+);diacetate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 222-164-4

RTECSNumber : AJ1900000

TSCA : TSCA

SDS Documents