missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium(II) 2,4-pentanedionate, Pd 34.7%

Carbonylation, oligomerization

Manufacturer:  Alfa Aesar™ 1051706

 View more versions of this product

Catalog No. AA1051706



Specifications

Specifications

Palladium(II) 2,4-pentanedionate
C10H14O4Pd
palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
JKDRQYIYVJVOPF-SYWGCQIGSA-L
(E)-4-oxopent-2-en-2-olate;palladium(2+)
53384484
Powder
Odorless
210°C (decomposition)
14024-61-4
MFCD00000025
Soluble in benzene and chloroform
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Pd+2]
304.638
304.92
Pd 34.7%
Yellow to Orange
5g

Chemical Identifiers

14024-61-4
304.638
JKDRQYIYVJVOPF-SYWGCQIGSA-L
53384484
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Pd+2]
C10H14O4Pd
MFCD00000025
palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
(E)-4-oxopent-2-en-2-olate;palladium(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

TSCA : TSCA

Documents